Assay Details
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869022.2 | XP_004869079.1 | 6 - 7 | 608 | 67 | |
XM_004869023.3 | XP_004869080.1 | 6 - 7 | 656 | 67 | ||
XM_021247779.1 | XP_021103438.1 | 6 - 7 | 631 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
