Assay Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854392.3 | XP_004854449.1 | 4 - 5 | 1066 | 92 | |
| XM_004854393.3 | XP_004854450.1 | 4 - 5 | 1067 | 92 | ||
| XM_004854394.3 | XP_004854451.1 | 4 - 5 | 1066 | 92 | ||
| XM_021266197.1 | XP_021121856.1 | 4 - 5 | 1069 | 92 | ||
| XM_021266198.1 | XP_021121857.1 | 4 - 5 | 1068 | 92 | ||
More Information
Set Membership: |
|

Csnk1a1
Genomic Map