Assay Details
| Gene Symbol: Pak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859093.2 | XP_004859150.1 | 5 - 6 | 1390 | 59 | |
| XM_004859094.2 | XP_004859151.1 | 4 - 5 | 937 | 59 | ||
| XM_013077550.2 | XP_012933004.1 | 5 - 6 | 1229 | 59 | ||
| XM_013077551.2 | XP_012933005.1 | 5 - 6 | 1268 | 59 | ||
| XM_021239679.1 | XP_021095338.1 | 5 - 6 | 1586 | 59 | ||
| XM_021239680.1 | XP_021095339.1 | 5 - 6 | 1523 | 59 | ||
| XM_021239681.1 | XP_021095340.1 | 4 - 5 | 1288 | 59 | ||
More Information
Set Membership: |
|

Pak4
Genomic Map