Assay Details
| Gene Symbol: Syncrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077952.2 | XP_012933406.2 | 3 - 4 | 494 | 100 | |
| XM_021240724.1 | XP_021096383.1 | 3 - 4 | 479 | 100 | ||
More Information
Set Membership: |
|

Syncrip
Genomic Map