Assay Details
| Gene Symbol: Vopp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013070417.2 | XP_012925871.1 | 3 - 4 | 322 | 67 | |
| XM_013070418.2 | XP_012925872.1 | 3 - 4 | 288 | 67 | ||
| XM_021254207.1 | XP_021109866.1 | 3 - 4 | 297 | 67 | ||
More Information
Set Membership: |
|

Vopp1
Genomic Map