Assay Details
| Gene Symbol: Slc35a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871963.3 | XP_004872020.1 | 3 - 4 | 642 | 78 | |
| XM_004871964.2 | XP_004872021.1 | 3 - 4 | 642 | 78 | ||
| XM_004871965.3 | XP_004872022.1 | 2 - 3 | 462 | 78 | ||
| XM_004871966.3 | XP_004872023.1 | 3 - 4 | 546 | 78 | ||
| XM_004871967.3 | XP_004872024.1 | 2 - 3 | 459 | 78 | ||
More Information
Set Membership: |
|

Slc35a2
Genomic Map