Assay Details
| Gene Symbol: Oaz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290044.1 | NP_001276973.1 | 4 - 5 | 655 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oaz2| Gene Symbol: Oaz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290044.1 | NP_001276973.1 | 4 - 5 | 655 | 76 | |
Set Membership: |
|