Assay Details
| Gene Symbol: Dhrs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870403.3 | XP_004870460.1 | 1 - 2 | 347 | 77 | |
| XM_021248673.1 | XP_021104332.1 | 1 - 2 | 365 | 77 | ||
| XM_021248674.1 | XP_021104333.1 | 1 - 2 | 351 | 77 | ||
More Information
Set Membership: |
|

Dhrs11
Genomic Map