Assay Details
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862254.2 | XP_004862311.1 | 3 - 4 | 267 | 63 | |
XM_004862255.2 | XP_004862312.1 | 3 - 4 | 402 | 63 | ||
XM_004862256.2 | XP_004862313.1 | 3 - 4 | 384 | 63 | ||
XM_021242237.1 | XP_021097896.1 | 3 - 4 | 276 | 63 | ||
XM_021242238.1 | XP_021097897.1 | 3 - 4 | 393 | 63 | ||
XM_021242239.1 | XP_021097898.1 | 3 - 4 | 411 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
