Assay Details
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285850.1 | NP_001272779.1 | 5 - 6 | 557 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
