Assay Details
| Gene Symbol: Rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310247.1 | NP_001297176.1 | 2 - 3 | 220 | 67 | |
| XM_021244625.1 | XP_021100284.1 | 2 - 3 | 235 | 67 | ||
| GenBank mRNA | KM486764.1 | - | 2 - 3 | 220 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021244625.1 transcripts.
Set Membership: |
|

Rgs5
Genomic Map