Assay Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310288.1 | NP_001297217.1 | 2 - 3 | 243 | 71 | |
| XM_021240527.1 | XP_021096186.1 | 3 - 4 | 298 | 71 | ||
| GenBank mRNA | KM486722.1 | - | 2 - 3 | 243 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021240527.1 transcripts.
Set Membership: |
|

Hprt1
Genomic Map