Assay Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310309.1 | NP_001297238.1 | 3 - 4 | 405 | 62 | |
| XM_021239197.1 | XP_021094856.1 | 3 - 4 | 275 | 62 | ||
| XM_021239198.1 | XP_021094857.1 | 4 - 5 | 516 | 62 | ||
| GenBank mRNA | KM486705.1 | - | 3 - 4 | 405 | 62 | |
More Information
Set Membership: |
|

Fech
Genomic Map