Assay Details
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301703.1 | NP_001288632.1 | 5 - 6 | 1127 | 68 | |
GenBank mRNA | HQ326690.2 | - | 5 - 6 | 1127 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
