Assay Details
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281982.1 | NP_001268911.1 | 1 - 2 | 81 | 100 | |
XM_021248968.1 | XP_021104627.1 | 1 - 2 | 478 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281982.1, XM_021248968.1 transcripts.
Set Membership: |
![]() ![]() |
