Assay Details
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281982.1 | NP_001268911.1 | 2 - 3 | 164 | 96 | |
XM_021248968.1 | XP_021104627.1 | 2 - 3 | 561 | 96 | ||
XM_021248969.1 | XP_021104628.1 | 2 - 3 | 105 | 96 | ||
XM_021248970.1 | XP_021104629.1 | 2 - 3 | 174 | 96 |
