Assay Details
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281982.1 | NP_001268911.1 | 4 - 5 | 404 | 66 | |
XM_021248968.1 | XP_021104627.1 | 4 - 5 | 801 | 66 | ||
XM_021248969.1 | XP_021104628.1 | 4 - 5 | 345 | 66 | ||
XM_021248970.1 | XP_021104629.1 | 4 - 5 | 414 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
