Assay Details
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001279838.1 | NP_001266767.1 | 2 - 3 | 308 | 88 | |
XM_021260469.1 | XP_021116128.1 | 2 - 3 | 598 | 88 | ||
GenBank mRNA | HQ326661.1 | - | 2 - 3 | 308 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
