Assay Details
Gene Symbol: Polr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001279871.1 | NP_001266800.1 | 3 - 4 | 370 | 61 | |
XM_013075570.2 | XP_012931024.1 | 3 - 4 | 451 | 61 | ||
XM_021265389.1 | XP_021121048.1 | 2 - 3 | 206 | 61 | ||
GenBank mRNA | HQ326674.2 | - | 3 - 4 | 371 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
