Assay Details
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310310.1 | NP_001297239.1 | 10 - 11 | 1127 | 63 | |
XM_013070578.2 | XP_012926032.1 | 9 - 10 | 960 | 63 | ||
XM_021254177.1 | XP_021109836.1 | 10 - 11 | 1094 | 63 | ||
GenBank mRNA | KM486682.1 | - | 10 - 11 | 1127 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
