Assay Details
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310258.1 | NP_001297187.1 | 7 - 8 | 1470 | 58 | |
XM_013068004.2 | XP_012923458.1 | 7 - 8 | 1513 | 58 | ||
XM_021249253.1 | XP_021104912.1 | 7 - 8 | 1085 | 58 | ||
GenBank mRNA | KM486735.1 | - | 7 - 8 | 1470 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
