Assay Details
Gene Symbol: LOC101701430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310234.1 | NP_001297163.1 | 1 | 118 | 93 | |
GenBank mRNA | LC012308.1 | - | 1 | 118 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
