Assay Details
Gene Symbol: LOC101708169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864145.3 | XP_004864202.1 | 6 - 7 | 766 | 98 | |
XM_021243878.1 | XP_021099537.1 | 6 - 7 | 781 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
