Assay Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858189.3 | XP_004858246.1 | 8 - 9 | 1210 | 62 | |
| XM_004858190.3 | XP_004858247.1 | 8 - 9 | 1210 | 62 | ||
| XM_013077132.2 | XP_012932586.1 | 8 - 9 | 1042 | 62 | ||
More Information
Set Membership: |
|

Hnf4a
Genomic Map