Assay Details
Gene Symbol: Cdkn2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865507.3 | XP_004865564.1 | 1 - 2 | 391 | 53 | |
XM_004865508.3 | XP_004865565.1 | 2 - 3 | 573 | 53 | ||
XM_004865509.3 | XP_004865566.1 | 1 - 2 | 391 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
