Assay Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853916.3 | XP_004853973.1 | 3 - 4 | 341 | 69 | |
XM_013075549.2 | XP_012931003.1 | 3 - 4 | 342 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
