Assay Details
| Gene Symbol: LOC101726496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866454.3 | XP_004866511.1 | 1 - 2 | 381 | 91 | |
| XM_013066101.2 | XP_012921555.1 | 1 - 2 | 383 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101726496| Gene Symbol: LOC101726496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866454.3 | XP_004866511.1 | 1 - 2 | 381 | 91 | |
| XM_013066101.2 | XP_012921555.1 | 1 - 2 | 383 | 91 | ||