Assay Details
Gene Symbol: LOC101726496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866454.3 | XP_004866511.1 | 4 - 5 | 837 | 105 | |
XM_013066101.2 | XP_012921555.1 | 4 - 5 | 839 | 105 |

More Information
Set Membership: |
![]() ![]() |
