Assay Details
| Gene Symbol: LOC101717359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836997.3 | XP_004837054.1 | 2 - 3 | 760 | 66 | |
| XM_013066639.2 | XP_012922093.1 | 2 - 3 | 657 | 66 | ||
More Information
Set Membership: |
|

LOC101717359
Genomic Map