Assay Details
Gene Symbol: LOC101717359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836997.3 | XP_004837054.1 | 3 - 4 | 1031 | 109 | |
XM_013066639.2 | XP_012922093.1 | 3 - 4 | 928 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
