Assay Details
| Gene Symbol: Carhsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864870.3 | XP_004864927.1 | 3 - 4 | 448 | 92 | |
| XM_004864871.3 | XP_004864928.1 | 3 - 4 | 382 | 92 | ||
| XM_013065649.2 | XP_012921103.1 | 3 - 4 | 848 | 92 | ||
| XM_013065650.2 | XP_012921104.1 | 3 - 4 | 431 | 92 | ||
| XM_021244495.1 | XP_021100154.1 | 3 - 4 | 519 | 92 | ||
More Information
Set Membership: |
|

Carhsp1
Genomic Map