Assay Details
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871046.3 | XP_004871103.1 | 6 - 7 | 1023 | 57 | |
XM_021249138.1 | XP_021104797.1 | 7 - 8 | 911 | 57 | ||
XM_021249139.1 | XP_021104798.1 | 6 - 7 | 1020 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
