Assay Details
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021262923.1 | XP_021118582.1 | 4 - 5 | 604 | 64 | |
XM_021262925.1 | XP_021118584.1 | 4 - 5 | 604 | 64 | ||
XM_021262926.1 | XP_021118585.1 | 4 - 5 | 382 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
