Assay Details
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068187.2 | XP_012923641.1 | 3 - 4 | 640 | 56 | |
XM_021249912.1 | XP_021105571.1 | 3 - 4 | 395 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021249912.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
