Assay Details
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021238718.1 | XP_021094377.1 | 2 - 3 | 443 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
