Assay Details
| Gene Symbol: Vdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855276.3 | XP_004855333.1 | 8 - 9 | 855 | 75 | |
| XM_004855277.3 | XP_004855334.1 | 9 - 10 | 833 | 75 | ||
| XM_004855278.3 | XP_004855335.1 | 7 - 8 | 850 | 75 | ||
| XM_021236742.1 | XP_021092401.1 | 8 - 9 | 791 | 75 | ||
| XM_021236743.1 | XP_021092402.1 | 7 - 8 | 781 | 75 | ||
| XM_021236744.1 | XP_021092403.1 | 8 - 9 | 764 | 75 | ||
| XM_021236745.1 | XP_021092404.1 | 8 - 9 | 830 | 75 | ||
| XM_021236746.1 | XP_021092405.1 | 7 - 8 | 754 | 75 | ||
More Information
Set Membership: |
|

Vdac3
Genomic Map