Assay Details
| Gene Symbol: Sugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068709.2 | XP_012924163.1 | 6 - 7 | 2579 | 54 | |
| XM_013068711.2 | XP_012924165.1 | 6 - 7 | 2532 | 54 | ||
| XM_021251020.1 | XP_021106679.1 | 6 - 7 | 2451 | 54 | ||
| XM_021251021.1 | XP_021106680.1 | 6 - 7 | 2503 | 54 | ||
More Information
Set Membership: |
|

Sugp2
Genomic Map