Assay Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853134.3 | XP_004853191.1 | 2 - 3 | 559 | 90 | |
| XM_004853135.3 | XP_004853192.1 | 2 - 3 | 559 | 90 | ||
| XM_013075288.2 | XP_012930742.1 | 3 - 4 | 621 | 90 | ||
More Information
Set Membership: |
|

Aqp4
Genomic Map