Assay Details
| Gene Symbol: Fgf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834770.3 | XP_004834827.1 | 2 - 3 | 967 | 63 | |
| XM_004834772.3 | XP_004834829.1 | 3 - 4 | 363 | 63 | ||
More Information
Set Membership: |
|

Fgf12
Genomic Map