Assay Details
Gene Symbol: Gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850107.2 | XP_004850164.1 | 4 - 5 | 744 | 64 | |
XM_021262642.1 | XP_021118301.1 | 4 - 5 | 1477 | 64 | ||
XM_021262643.1 | XP_021118302.1 | 4 - 5 | 1444 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
