Assay Details
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004868039.2 | XP_004868096.1 | 3 - 4 | 567 | 69 | |
XM_021247016.1 | XP_021102675.1 | 3 - 4 | 578 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
