Assay Details
| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856682.3 | XP_004856739.1 | 6 - 7 | 980 | 54 | |
| XM_004856684.3 | XP_004856741.1 | 4 - 5 | 571 | 54 | ||
More Information
Set Membership: |
|

Il10ra
Genomic Map