Assay Details
| Gene Symbol: LOC101700218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856420.3 | XP_004856477.2 | 5 - 6 | 1573 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101700218| Gene Symbol: LOC101700218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856420.3 | XP_004856477.2 | 5 - 6 | 1573 | 138 | |
Set Membership: |
|