Assay Details
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839598.3 | XP_004839655.1 | 7 - 8 | 1062 | 65 | |
XM_021254153.1 | XP_021109812.1 | 8 - 9 | 1188 | 65 | ||
XM_021254154.1 | XP_021109813.1 | 7 - 8 | 915 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
