Assay Details
Gene Symbol: Atp6v1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869950.3 | XP_004870007.1 | 6 - 7 | 715 | 60 | |
XM_004869951.3 | XP_004870008.1 | 6 - 7 | 613 | 60 | ||
XM_004869952.3 | XP_004870009.1 | 6 - 7 | 589 | 60 | ||
XM_021248370.1 | XP_021104029.1 | 6 - 7 | 613 | 60 | ||
XM_021248371.1 | XP_021104030.1 | 5 - 6 | 466 | 60 | ||
XM_021248372.1 | XP_021104031.1 | 6 - 7 | 613 | 60 | ||
XM_021248373.1 | XP_021104032.1 | 6 - 7 | 613 | 60 | ||
XM_021248374.1 | XP_021104033.1 | 5 - 6 | 466 | 60 | ||
XM_021248375.1 | XP_021104034.1 | 6 - 7 | 613 | 60 | ||
XM_021248376.1 | XP_021104035.1 | 5 - 6 | 635 | 60 | ||
XM_021248377.1 | XP_021104036.1 | 6 - 7 | 613 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
