Assay Details
Gene Symbol: Lgi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838530.3 | XP_004838587.1 | 6 - 7 | 985 | 85 | |
XM_013069862.2 | XP_012925316.1 | 4 - 5 | 843 | 85 | ||
XM_013069863.2 | XP_012925317.1 | 6 - 7 | 980 | 85 | ||
XM_021253071.1 | XP_021108730.1 | 5 - 6 | 915 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
