Assay Details
Gene Symbol: Lrfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858955.3 | XP_004859012.1 | 1 - 2 | 1048 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004858955.3 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
