Assay Details
Gene Symbol: LOC101707360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850338.2 | XP_004850395.1 | 15 - 16 | 2419 | 69 | |
XM_004850339.2 | XP_004850396.1 | 14 - 15 | 2359 | 69 | ||
XM_021262849.1 | XP_021118508.1 | 15 - 16 | 2498 | 69 | ||
XM_021262850.1 | XP_021118509.1 | 15 - 16 | 2495 | 69 | ||
XM_021262851.1 | XP_021118510.1 | 14 - 15 | 2438 | 69 | ||
XM_021262852.1 | XP_021118511.1 | 14 - 15 | 2435 | 69 | ||
XM_021262853.1 | XP_021118512.1 | 15 - 16 | 2498 | 69 | ||
XM_021262854.1 | XP_021118513.1 | 15 - 16 | 2498 | 69 | ||
XM_021262855.1 | XP_021118514.1 | 15 - 16 | 2498 | 69 | ||
XM_021262856.1 | XP_021118515.1 | 8 - 9 | 943 | 69 | ||
XM_021262857.1 | XP_021118516.1 | 1 - 2 | 124 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021262857.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
