Assay Details
Gene Symbol: LOC101707360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850338.2 | XP_004850395.1 | 19 - 20 | 2937 | 78 | |
XM_004850339.2 | XP_004850396.1 | 18 - 19 | 2877 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
