Assay Details
Gene Symbol: LOC101707360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850338.2 | XP_004850395.1 | 5 - 6 | 465 | 95 | |
XM_004850339.2 | XP_004850396.1 | 5 - 6 | 465 | 95 | ||
XM_021262849.1 | XP_021118508.1 | 5 - 6 | 541 | 95 | ||
XM_021262850.1 | XP_021118509.1 | 5 - 6 | 541 | 95 | ||
XM_021262851.1 | XP_021118510.1 | 5 - 6 | 541 | 95 | ||
XM_021262852.1 | XP_021118511.1 | 5 - 6 | 541 | 95 | ||
XM_021262853.1 | XP_021118512.1 | 5 - 6 | 541 | 95 | ||
XM_021262854.1 | XP_021118513.1 | 5 - 6 | 541 | 95 | ||
XM_021262855.1 | XP_021118514.1 | 5 - 6 | 541 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
